Learn More
Tocris Bioscience™ Abirateronacetat
Beschreibung
Abiraterone acetate is a potent CYP17A1 inhibitor (IC50 values are 17 nM and 18 nM for 17,20-lyase activity and 17α-hydroxylase activity, respectively). Inhibits intratumor androgen synthesis in prostate cancer.
Spezifikation
Spezifikation
| Chemischer Name oder Material | (3β)-17-(3-Pyridinyl)androsta-5,16-dien-3-yl acetate |
| CAS | 154229-18-2 |
| Menge | 5 mg |
| Summenformel | C26H33NO2 |
| Reinheit | 98% |