Learn More
ABRA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA558042
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: CNYNHHLDVV DNLTLMVEHT DIPEASPAST PQIIKHKTLD LDDRWQFKRS RLLDTQDKRS KADTGSSNQD KASKMSSPET DEEIEKMKGF GEYSRSPT Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 50%.
ABRA1 Polyclonal antibody specifically detects ABRA1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ABRA1 | |
Polyclonal | |
Unconjugated | |
Abraxas1 | |
3830405G04Rik; 5630400M01Rik; ABRA1; abraxas protein; Abraxas1; AI506069; AL024423; AV118690; BRCA1-A complex subunit Abraxas; BRCA1-A complex subunit Abraxas 1; Ccdc98; coiled-coil domain containing 98; coiled-coil domain-containing protein 98; FAM175A; family with sequence similarity 175 member A; family with sequence similarity 175, member A; FLJ11520; FLJ12642; FLJ13614; Protein FAM175A; RGD1305287; UNQ496/PRO1013 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84142 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWZ7 | |
Abraxas1 | |
Recombinant protein corresponding to Human ABRA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only