missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACAA2 Polyclonal Antibody

Description
Positive Control: HeLa Predicted Reactivity: Rat (90%), Zebrafish (80%), Pig (93%), Chicken (83%), Rhesus Monkey (98%), Bovine (92%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ACAA2 (3-Ketoacyl-CoA thiolase) is an enzyme that in humans is encoded by the ACAA2 gene. Acetyl-Coenzyme A acyltransferase 2 is an acetyl-CoA C-acyltransferase enzyme. The encoded protein catalyzes the last step of the mitochondrial fatty acid beta-oxidation spiral. Unlike most mitochondrial matrix proteins, it contains a non-cleavable amino-terminal targeting signal.
Specifications
Specifications
| Antigen | ACAA2 |
| Anwendungen | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Konzentration | 1.01 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gen | ACAA2 |
| Gen-Zugriffsnummer | P42765, Q8BWT1 |
| Gen-Alias | 0610011L04Rik; 3-ketoacyl-CoA thiolase, mitochondrial; Acaa2; Acetyl-CoA acetyltransferase; Acetyl-CoA acyltransferase; acetyl-CoA acyltransferase 2; acetyl-Coenzyme A acyltransferase 2; acetyl-Coenzyme A acyltransferase 2 (mitochondrial 3-oxoacyl-Coenzyme A thiolase); Acyl-CoA hydrolase, mitochondrial; AI255831; AI265397; beta ketothiolase; beta-ketothiolase; D18Ertd240e; DSAEC; mitochondrial 3-oxoacyl-CoA thiolase; mitochondrial 3-oxoacyl-Coenzyme A thiolase; T1 |
| Gensymbole | ACAA2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?