missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAD11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550295
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total ACAD11 protein.
ACAD11 Polyclonal antibody specifically detects ACAD11 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ACAD11 | |
Polyclonal | |
Unconjugated | |
ACAD11 | |
62113 protein; ACAD11; ACAD-11; Acyl-CoA dehydrogenase family member 11; acyl-CoA dehydrogenase family, member 11; acyl-Coenzyme A dehydrogenase family, member 11; FLJ12592; MGC150619 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84129 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q709F0 | |
ACAD11 | |
Fusion protein corresponding to a region derived from internal residues of human Acyl-CoA dehydrogenase family, member 11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |