Learn More
ACAD8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560547
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LSFGKKEKKV GWNSQPTRAV IFEDCAVPVA NRIGSEGQGF LIAVRGLNGG RINIASCSLG AAHASVILTR DHLNVRKQFG EPLASNQYLQ FTLADMA Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 47%.
This gene encodes a member of the acyl-CoA dehydrogenase family of enzymes that catalyze the dehydrogenation of acyl-CoA derivatives in the metabolism of fatty acids or branch chained amino acids. The encoded protein is a mitochondrial enzyme that functions in catabolism of the branched-chain amino acid valine. Defects in this gene are the cause of isobutyryl-CoA dehydrogenase deficiency.Spezifikation
ACAD8 | |
Polyclonal | |
Unconjugated | |
ACAD8 | |
2310016C19Rik; Acad8; ACAD-8; Activator-recruited cofactor 42 kDa component; acyl-CoA dehydrogenase family member 8; acyl-CoA dehydrogenase family, member 8; acyl-Coenzyme A dehydrogenase family, member 8; AI786953; ARC42; IBD; IBDH; Isobutyryl-CoA dehydrogenase, mitochondrial; RGD1564209 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27034 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKU7 | |
ACAD8 | |
Recombinant protein corresponding to Human ACAD8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only