missing translation for 'onlineSavingsMsg'
Learn More

ACAD8 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA559250

Weitere Details : Netto-Gewicht : 0.02000kg

Artikelnummer. 15747111

  • 514.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Immunogen sequence: ELFPVDVMRK AAQLGFGGVY IQTDVGGSGL SRLDTSVIFE ALATGCTSTT AYISIHNMCA WMIDSFGNEE QRHKFCPPL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

ACAD8 Polyclonal antibody specifically detects ACAD8 in Human samples. It is validated for Immunohistochemistry.
Spezifikation

Spezifikation

ACAD8
Polyclonal
Unconjugated
ACAD8
2310016C19Rik; Acad8; ACAD-8; Activator-recruited cofactor 42 kDa component; acyl-CoA dehydrogenase family member 8; acyl-CoA dehydrogenase family, member 8; acyl-Coenzyme A dehydrogenase family, member 8; AI786953; ARC42; IBD; IBDH; Isobutyryl-CoA dehydrogenase, mitochondrial; RGD1564209
Rabbit
Antigen affinity chromatography
RUO
27034
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9UKU7
ACAD8
Recombinant protein corresponding to Human ACAD8
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only