missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAD9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550294
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total ACAD9 protein.
ACAD9 Polyclonal antibody specifically detects ACAD9 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
ACAD9 | |
Polyclonal | |
Unconjugated | |
Acad9 | |
2600017P15Rik; 4732402K02; ACAD9; ACAD-9; Acyl-CoA dehydrogenase family member 9; acyl-CoA dehydrogenase family member 9, mitochondrial; acyl-CoA dehydrogenase family, member 9; acyl-Coenzyme A dehydrogenase family, member 9; C630012L17Rik; Complex I assembly factor ACAD9, mitochondrial; NPD002; Nyggf2; very-long-chain acyl-CoA dehydrogenase VLCAD; very-long-chain acyl-CoA dehydrogenase VLCAD homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28976 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9H845 | |
Acad9 | |
Fusion protein corresponding to residues near the C terminal of human acyl-CoA dehydrogenase family, member 9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |