missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACADVL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14527-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This antibody is specific to ACADVL.
The protein encoded by this gene is targeted to the inner mitochondrial membrane where it catalyzes the first step of the mitochondrial fatty acid beta-oxidation pathway. This acyl-Coenzyme A dehydrogenase is specific to long-chain and very-long-chain fatty acids. A deficiency in this gene product reduces myocardial fatty acid beta-oxidation and is associated with cardiomyopathy. Alternative splicing results in multiple transcript variants encoding different isoforms.Spezifikation
ACADVL | |
Polyclonal | |
Unconjugated | |
ACADVL | |
ACAD6, ACADVL, LCACD, VLCAD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11370, 25363, 37 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P49748, P50544 | |
ACADVL | |
ACADVL Fusion Protein Ag6016 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |