Learn More
ACCS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554485
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NVVVLNGGAS LFSALATVLC EAGEAFLIPT PYYGAITQHV CLYGNIRLAY VYLDSEVTGL DTRPFQLTVE KLEMALREAH SEGVKVKGLI LISP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
ACCS Polyclonal antibody specifically detects ACCS in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
ACCS | |
Polyclonal | |
Unconjugated | |
ACCS | |
1-aminocyclopropane-1-carboxylate synthase (non-functional); 1-aminocyclopropane-1-carboxylate synthase homolog; 1-aminocyclopropane-1-carboxylate synthase homolog (Arabidopsis)(non-functional); 1-aminocyclopropane-1-carboxylate synthase homolog (inactive); 1-aminocyclopropane-1-carboxylate synthase-like protein (non-functional) isoform 1; 1-aminocyclopropane-1-carboxylate synthase-like protein (non-functional) isoform 2; 1-aminocyclopropane-1-carboxylate synthase-like protein (non-functional) isoform 4; 1-aminocyclopropane-1-carboxylate synthase-like protein 1; 2610203E10Rik; ACC synthase-like protein 1; ACCS; ACS; Phacs; RGD1309314 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84680 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96QU6 | |
ACCS | |
Recombinant protein corresponding to Human ACCS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only