Learn More
Acetic Acid D4 99.91% D, Eurisotop
Description
Acetic Acid D4
- Isotopic Enrichment: 99.91% D
- Quantity: (2) 0.75 mL
- Packaging: Ampule
- Water Content: water <0.05%
CoA and MSDS available at Eurisotop.com
Specifications
Specifications
| Color | Colorless |
| Deuterium (D) | 99.91% D |
| Packaging | Ampule |
| Quantity | 2 x 0.75 mL |
| CAS | 1186-52-3 |
| Melting Point | 16.7°C |
| Boiling Point | 118°C |
| Molecular Formula | C2H4O2 |
| Synonym | Acetic-d3 acid-d, Tetradeuteroacetic acid, Acetic-d4 |
| InChI Key | QTBSBXVTEAMEQO-GUEYOVJQSA-N |
| Show More |