Learn More
H2BK12ac Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke:  Invitrogen PA540100
Weitere Details : Netto-Gewicht : 0.00900kg
Artikelnummer. 15526791
  • 471.00 € / 50 Mikrogramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
H2BK12ac Polyclonal antibody specifically detects H2BK12ac in Human samples. It is validated for ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Immunocytochemistry, Peptide Array, Western Blot.
Spezifikation
Spezifikation
H2BK12ac
Polyclonal
Unconjugated
H2BC21
2610022J01Rik; Acetyl-Histone H2; Acetyl-Histone H2B K12; Acetyl-Histone H2B Lys12; AV127319; bA317E16.3; dJ160A22.3; dJ193B12.2; dJ221C16.3; dJ221C16.6; dJ221C16.8; dJ97D16.4; GL105; H2B; H2B 291A; H2B 291B; H2b 613; H2B GL105; H2B histone family member M; H2B histone family member W testis-specific; H2B histone family member W, testis specific; H2B histone family, member A; H2B histone family, member B; H2B histone family, member C; H2B histone family, member D; H2B histone family, member E; H2B histone family, member F; H2B histone family, member J; H2B histone family, member K; H2B histone family, member L; H2B histone family, member M; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member R; H2B histone family, member S; H2B histone family, member T; H2B histone family, member U, (testis-specific); H2B histone family, member W, testis-specific; H2B K; H2B type 12; H2B.1; H2B.1A; H2B.1B; H2B.2; H2B/a; H2B/b; H2B/c; H2B/d; H2B/e; H2B/f; H2B/g; H2B/h;
Rabbit
Antigen affinity chromatography
RUO
102724334, 114483833, 128312, 158983, 255626, 286436, 3017, 3018, 440689, 8339, 8340, 8341, 8342, 8345, 8346, 8347, 8348, 8349, 85236, 8970
-20°C or -80°C if preferred
Liquid
ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Immunocytochemistry, Peptide Array, Western Blot
0.82 mg/mL
PBS with 0.05% ProClin 300, 0.05% sodium azide; pH 7.4
A0A2R8Y619, O60814, P06899, P0C1H6, P23527, P33778, P57053, P58876, P62807, Q16778, Q5QNW6, Q7Z2G1, Q8N257, Q93079, Q96A08, Q99877, Q99879, Q99880
H2B, H2bc1, H2BC10, H2BC11, H2BC12, H2BC13, H2BC14, H2BC15, H2BC17, H2bc18, H2BC21, H2BC3, H2BC4, H2BC5, H2BC8, H2BC9, H2BE1, H2BU1, H2BW1, H2BW2, LOC102724334
KLH-conjugated synthetic peptide corresponding to human the region of histone H2B containing the acetylated lysine 12 (H2BK12ac).