Learn More
H3K79ac Recombinant Rabbit Monoclonal Antibody (RM156), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen MA524675
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody reacts to Histone H3 acetylated at Lysine 79 (K79ac). No cross reactivity with other acetylated Lysines in Histone H3. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
H3K79ac Recombinant Monoclonal antibody specifically detects H3K79ac in Human samples. It is validated for ChIP Assay, ELISA, Immunocytochemistry, Luminex, Western Blot.Spezifikation
H3K79ac | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH 7.2-7.4 | |
P68431, Q71DI3 | |
H3C1, H3C10, H3C12, H3C14, H3C15, H3C2, H3C7, HIST1H3A | |
Acetyl-peptide corresponding to the Acetyl-Histone H3 (Lys79). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ChIP Assay, ELISA, Immunocytochemistry, Luminex, Western Blot | |
RM156 | |
Unconjugated | |
H3C1 | |
Acetyl H3; Acetyl Histone H3; Acetyl-H3 K79; Acetyl-H3 Lys79; Acetyl-Histone H3 K79; Acetyl-Histone H3 Lys79; H3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3C; H3 K79ac; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; H3.3A; H3.3B; H3.5; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-143; H3-291; H3-3A; H3-3B; H3-53; H3-614; H3a; H3b; H3-B; H3C1; H3c10; H3C11; H3C12; H3C2; H3C3; H3C4; H3C6; H3C7; H3c8; H3f; H3-F; H3F1K; H3F2; H3F3; H3F3A; H3F3B; H3F3C; H3FA; H3FB; H3FC HIST1H3C; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; H3FM; H3FN; H3g; H3h; H3i; H3K79; H3Lys79ac; Hist1; HIST1H3A; Hist1h3b; Hist1h3c; HIST1H3D; Hist1h3e; Hist1h3f; HIST1H3G; Hist1h3h; Hist1h3i; HIST1H3J; HIST2H3A; Hist2h3b; HIST2H3C; Hist2h3c1; | |
Rabbit | |
Protein A | |
RUO | |
126961, 333932, 8350, 8356, 8357, 8358, 8968 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
For Research Use Only