missing translation for 'onlineSavingsMsg'
Learn More

H3K9ac Monoclonal Antibody, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA523517

Weitere Details : Netto-Gewicht : 0.00900kg

Artikelnummer. 15537281

  • 469.00 € / 50 Mikrogramm
Verfügbar ab: 02-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

H3K9ac Monoclonal antibody specifically detects H3K9ac in Human, Mouse samples. It is validated for Immunocytochemistry, Peptide Array, Western Blot.
Spezifikation

Spezifikation

H3K9ac
Monoclonal
Unconjugated
H3C15
Acetyl H3; Acetyl Histone H3; Acetyl-H3 K9; Acetyl-H3 Lys9; Acetyl-Histone H3 K9; Acetyl-Histone H3 Lys9; B0035.7; BUR5; CELE_T10C6.12; CG31613; CG31613-PA; CG33803; CG33806; CG33809; CG33812; CG33815; CG33818; CG33821; CG33824; CG33827; CG33830; CG33833; CG33836; CG33839; CG33842; CG33845; CG33848; CG33851; CG33854; CG33857; CG33860; CG33863; CG33866; Dmel\CG31613; Dmel_CG31613; F07B7.10; F07B7.3; F08G2.2; F17E9.13; F35H10.1; F45F2.4; F54E12.5; F55G1.10; H02I12.7; H3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3B.1; H3 histone, family 3C; H3 K9ac; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; h3.2a; H3.3A; H3.3B; H3.5; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-143; H3-291; H3-3A; H3-3B; h3-5; H3-53; H3-
Mouse
Protein A
RUO
126961, 260423, 319150, 319152, 333932, 360198, 8350, 8356, 8357, 8358, 8968, 97114
-20°C or -80°C if preferred
Liquid
Immunocytochemistry, Peptide Array, Western Blot, Western Blot
1 mg/mL
PBS with 0.05% sodium azide; pH 7.4
P68431, P68433, P84228, Q71DI3
H3C1, H3C10, H3C12, H3C14, H3C15, H3C2, H3C7
KLH-conjugated synthetic peptide corresponding to human histone H3 acetylated at lysine 9 (H3K9ac).
50 μg
Primary
Human, Mouse
Antibody
IgG2b
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote