missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACKR2 Mouse anti-Human, Clone: 1A3A2, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 60046-1-IG-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a beta chemokine receptor, which is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptor-mediated signal transduction are critical for the recruitment of effector immune cells to the inflammation site. This gene is expressed in a range of tissues and hemopoietic cells. The expression of this receptor in lymphatic endothelial cells and overexpression in vascular tumors suggested its function in chemokine-driven recirculation of leukocytes and possible chemokine effects on the development and growth of vascular tumors. This receptor appears to bind the majority of beta-chemokine family members; however, its specific function remains unknown. This gene is mapped to chromosome 3p21.3, a region that includes a cluster of chemokine receptor genes.Spezifikation
ACKR2 | |
Monoclonal | |
0.83 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O00590 | |
ACKR2 | |
ACKR2 Fusion Protein Ag1941 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin) | |
1A3A2 | |
Unconjugated | |
ACKR2 | |
ACKR2, C C chemokine receptor D6, CCBP2, CCR10, CCR9, chemokine binding protein 2, Chemokine binding protein D6, Chemokine receptor CCR 10, Chemokine receptor CCR 9, CMKBR9, D6, hD6 | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
1238 | |
-20°C | |
Liquid |