missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT7L Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 51016-2-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Spezifikation
ACOT7L | |
Polyclonal | |
Unconjugated | |
LOC344967 | |
LOC344967 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BACHL, ACOT7L, LOC344967 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
344967 | |
-20°C | |
Liquid |