missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550303
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total ACOT9 protein.
ACOT9 Polyclonal antibody specifically detects ACOT9 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ACOT9 | |
Polyclonal | |
Unconjugated | |
ACOT9 | |
0610041P13Rik; Acate2; ACOT9; Acyl coenzyme A thioester hydrolase 2; acyl-CoA thioester hydrolase 9; Acyl-CoA thioesterase 9; acyl-Coenzyme A thioesterase 2, mitochondrial; acyl-coenzyme A thioesterase 9, mitochondrial; C76421; CGI-16; Mitochondrial 48 kDa acyl-CoA thioester hydrolase 1; mitochondrial acyl-CoA thioesterase; MTACT48; MT-ACT48; mt-ACT48.1; MTE-2; p48; Protein U8; U8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23597 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9Y305 | |
ACOT9 | |
Full length fusion protein of Human ACOT9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |