missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550297
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total ACOX2 protein.
ACOX2 Polyclonal antibody specifically detects ACOX2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ACOX2 | |
Polyclonal | |
Unconjugated | |
ACOX2 | |
3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholestanoyl-CoA 24-hydroxylase; 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholestanoyl-CoA oxidase; Acox2; acyl-CoA oxidase 2; acyl-CoA oxidase 2, branched chain; acyl-Coenzyme A oxidase 2, branched chain; BCOX; branched chain acyl-CoA oxidase; BRCACOX; BRCOX; Peroxisomal acyl-coenzyme A oxidase 2; peroxisomal branched chain acyl-CoA oxidase; THCA-CoA oxidase; THCCox; trihydroxycoprostanoyl-CoA oxidase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8309, 93732 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q99424, Q9QXD1 | |
ACOX2 | |
Fusion protein corresponding to residues near the C terminal of human Acyl-CoA oxidase 2, branched chain | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |