missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ACP2 (Human) Recombinant Protein
Human ACP2 full-length ORF ( AAH03160, 1 a.a. - 423 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00000053-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: MAGKRSGWSRAALLQLLLGVNLVVMPPTQARSLRFVTLLYRHGDRSPVKTYPKDPYQEEEWPQGFGQLTKEGMLQHWELGQALRQRYHGFLNTSYHRQEVYVRSTDFDRTLMSAEANLAGLFPPNGMQRFNPNISWQPIPVHTVPITEDRLLKFPLGPCPRYEQLQNETRQTPEYQNESSRNAQFLDMVANETGLTDLTLETVWNVYDTLFCEQTHGLRLPPWASPQTMQRLSRLKDFSFRFLFGIYQQAEKARLQGGVLLAQIRKNLTLMATTSQLPKLLVYSAHDTTLVALQMALDVYNGEQAPYASCHIFELYQEDSGNFSVEMYFRNESDKAPWPLSLPGCPHRCPLQDFLRLTEPVVPKDWQQECQLASGPADTEVIVALAVCGSILFLLIVLLLTVLFRMQAQPPGYRHVADGEDHA
Spezifikation
AAH03160 | |
acid phosphatase 2, lysosomal | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
53 | |
Wheat germ expression system | |
25 μg | |
ACP2 | |
GST |