missing translation for 'onlineSavingsMsg'
Learn More

ACP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 15236-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16884324

  • 329.85 € / 150 Mikroliter

Voraussichtliches Versanddatum: 18-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene encodes the beta subunit of lysosomal acid phosphatase (LAP). LAP is chemically and genetically distinct from red cell acid phosphatase. The encoded protein belongs to a family of distinct isoenzymes which hydrolyze orthophosphoric monoesters to alcohol and phosphate. Mutations in this gene or in the related alpha subunit gene cause acid phosphatase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
Spezifikation

Spezifikation

ACP2
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P11117, P20611, P24638
acid phosphatase 2, lysosomal, ACP2, LAP, Lysosomal acid phosphatase
Rabbit
IgG
150 μL
-20°C
Primary
11432, 24162, 53
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
ACP2
Liquid
ACP2
ACP2 Fusion Protein Ag7356
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation