missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17521-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This antibody is specific to the ACTA1. It does not cross-react with other actin isoforms.
The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.Spezifikation
ACTA1 | |
Polyclonal | |
Unconjugated | |
ACTA1 | |
ACTA, ACTA1, ACTA1 skeletal Muscle Actin, ACTA1 skeletal Muscle Actin Specific, Actin, alpha skeletal muscle, Alpha actin 1, ASMA, CFTD, CFTD1, CFTDM, MPFD, NEM1, NEM2, NEM3, sarcomeric actin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11459, 29437, 58 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P68133, P68134, P68136 | |
Acta1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |