Learn More
ACROBiosystems ActiveMax™ Human SCF / KITLG Protein, Tag Free
N.V.
Spezifikation
Zugriffsnummer | AAH69797 |
---|---|
Molekulargewicht | 22.1 kDa |
Quelle | ActiveMax™ Human SCF, Tag Free (SCF-H4212) is expressed from human 293 cells (HEK293). It contains AA Glu 26 - His 214 (Accession # AAH69797). |
Immunogen | Glu 26 - His 214 |
Lagerungsbedingungen | -20°C |
Beschreibung
Stem Cell Factor is also known as SCF, kit-ligand, KL, steel factor, KITLG, FPH2, KL-1, Kitl, MGF, SCF, SF, or SHEP7, and is a cytokine that binds to the c-Kit receptor (CD117). SCF can exist both as a transmembrane protein and a soluble protein. This cytokine plays an important role in hematopoiesis (formation of blood cells), spermatogenesis, and melanogenesis. The soluble and transmembrane forms of the protein are formed by alternative splicing of the same RNA transcript.Soluble and transmembrane SCF is produced by fibroblasts and endothelial cells. Soluble SCF has a molecular weight of 18,5 KDa and forms a dimer. SCF plays an important role in the hematopoiesis during embryonic development. Sites where hematopoiesis takes place, such as the fetal liver and bone marrow, all express SCF. During development, the presence of the SCF also plays an important role in the localization of melanocytes, cells that produce melanin and control pigmentation. SCF plays a role in the regulation of HSCs in the stem cell niche in the bone marrow. SCF may be used along with other cytokines to culture HSCs and hematopoietic progenitors. The expansion of these cells ex-vivo (outside the body) would allow advances in bone-marrow transplantation, in which HSCs are transferred to a patient to re-establish blood formation.
This protein carries no “tag”. The protein has a calculated MW of 22.1 kDa. The protein migrates as 32-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH69797 | |
ActiveMax™ Human SCF, Tag Free (SCF-H4212) is expressed from human 293 cells (HEK293). It contains AA Glu 26 - His 214 (Accession # AAH69797). | |
-20°C | |
SCF | |
Human | |
PBS, pH7.4 | |
HEK293 | |
ActiveMax™ Cytokines |
22.1 kDa | |
Glu 26 - His 214 | |
KITLG,FPH2,KL-1,Kitl,MGF,SCF,SF,SHEP7,KL | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>98% as determined by SDS-PAGE |