Learn More
ACYP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557216
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EVFGRVQGVC FRMYTEDEAR KIGVVGWVKN TSKGTVTGQV QGPEDKVNSM KSWLSKVGSP SSRIDRTNFS NE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.
ACYP2 Polyclonal antibody specifically detects ACYP2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ACYP2 | |
Polyclonal | |
Unconjugated | |
ACYP2 | |
2310004B09Rik; acylphosphatase 2; acylphosphatase 2, muscle type; acylphosphatase, muscle type isozyme; Acylphosphatase-2; acylphosphate phosphohydrolase 2; ACYM; ACYP; ACYP2; testicular tissue protein Li 11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
98 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P14621 | |
ACYP2 | |
Recombinant protein corresponding to Human ACYP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only