missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAR1 Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14330-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes the enzyme responsible for RNA editing by site-specific deamination of adenosines. This enzyme destabilizes double-stranded RNA through conversion of adenosine to inosine. Mutations in this gene have been associated with dyschromatosis symmetrica hereditaria. Alternative splicing results in multiple transcript variants.Spezifikation
ADAR1 | |
Polyclonal | |
Unconjugated | |
ADAR | |
ADAR, ADAR1, adenosine deaminase, RNA, DRADA, DSH, DSRAD, G1P1, IFI 4, IFI4, Interferon inducible protein 4, K88dsRBP, p136 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100523784, 103, 56417, 81635 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P55265, P55266, Q99MU3 | |
Adar | |
ADAR1 Fusion Protein Ag5609 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG |