missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15133-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes an enzyme that belongs to the acireductone dioxygenase family of metal-binding enzymes, which are involved in methionine salvage. This enzyme may regulate mRNA processing in the nucleus, and may carry out different functions depending on its localization. A pseudogene that is located on chromosome 20 has been defined for this gene.Spezifikation
ADI1 | |
Polyclonal | |
Unconjugated | |
ADI1 | |
Acireductone dioxygenase, acireductone dioxygenase 1, ADI1, APL1, ARD, HMFT1638, MTCBP 1, MTCBP1, SIP L, SIPL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
104923, 298934, 55256 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q562C9, Q99JT9, Q9BV57 | |
ADI1 | |
ADI1 Fusion Protein Ag7274 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |