missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Heavy duty lab models with stainless steel interior:
- Delivery to final end-user location
- Including unpacking and packaging disposal
Specifications
Specifications
| For Use With (Equipment) | Heavy duty/ Steel inner liner |
| Product Type | Advanced delivery |
| Duration | 2 Weeks |
| Target Audience | Research Laboratories |
| Species | All |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?