missing translation for 'onlineSavingsMsg'
Learn More

Liebherr™ Advanced Delivery Service ULT Models

Product Code. 30281198 Shop All Liebherr Appliances Products
Change view
Click to view available options
Duration:
2 Weeks
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Duration Unit Size
30281198 2 Weeks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30281198 Supplier Liebherr™ Supplier No. Delivery_Adv_ULT
 Restricted Item
Verfügbar ab: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Advanced Delivery Service ULT Models to final end-user location.

  • ULT models:
  • Delivery to final end-user location
  • Including unpacking and packaging disposal

Specifications

For Use With (Equipment) ULT
Product Type Advanced delivery
Duration 2 Weeks
Target Audience Research Laboratories
Species All
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.