missing translation for 'onlineSavingsMsg'
Learn More

Liebherr™ Advanced Delivery Service Upright Lab Models

Product Code. 30281199 Shop All Liebherr Appliances Products
Change view
Click to view available options
Duration:
2 Weeks
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Duration Unit Size
30281199 2 Weeks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30281199 Supplier Liebherr™ Supplier No. Delivery_Adv_Standgeräte
 Restricted Item
Verfügbar ab: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Advanced Delivery Service Upright Lab Models to final end-user location.

  • Upright lab models with polystyrol interior:
  • Delivery to final end-user location
  • Including unpacking and packaging disposal

Specifications

For Use With (Equipment) Uprights
Duration 2 Weeks
Product Type Advanced delivery
Target Audience Research Laboratories
Species Human
Product Title
Select an issue

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.