missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clinitrol™ 290 Reference Solution, 5mL; Advanced Instruments
Critical normal human serum midpoint of 290mOsm
Marke: Advanced Instruments AI-3LA029
Weitere Details : Netto-Gewicht : 0.05000kg
Beschreibung
Critical normal human serum midpoint of 290mOsm
- Checks performance of freezing-point osmometers in the critical area of normal serum values
- Contains known and controlled quantities of some of the same solutes which comprise osmotically active particles of serum
- Pipetted into a tube and run just like normal serum
- Runs as the first and last samples in a series of tests to monitor operator technique and instrument performance
- Does not require mixing, reconstituting or refrigerating; one-year shelf life
Spezifikation
CE | |
2°C to 30°C | |
Sodium Chloride (NaCl) and 2-Hydroxybiphenyl Sodium Salt Tetrahydrate | |
10 x 5 mL |
Clear | |
Serum | |
Osmometer instrument calibration and quality control |