Learn More
Tocris Bioscience™ AEG 40730 dihydrochloride
IAP antagonist; induces apoptosis
N.V.
Beschreibung
Beschreibung
IAP antagonist; binds to the BIR3 domain of cIAP1, cIAP2 and XIAP with nanomolar affinity. Reduces cIAP1, cIAP2 and XIAP protein levels in human breast cancer MDA-MB-231 cells. Induces apoptosis in combination with TNF, and potentiates TRAIL-mediated apoptosis in human colorectal carcinoma HCT 116 cells. Cell permeable.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Produktzertifizierungen
For Research Use Only