missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AF-6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 55102-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The antibody is specific to MLLT4.
AFDN belongs to an adhesion system, probably together with the E-cadherin-catenin system, which plays a role in the organization of homotypic, interneuronal and heterotypic cell-cell adherens junctions (AJs). Nectin- and actin-filament-binding protein that connects nectin to the actin cytoskeleton.Spezifikation
AF-6 | |
Polyclonal | |
Unconjugated | |
AFDN | |
AF 6, AF6, AFADIN, Afadin variant, MLLT4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17356, 26955, 4301 | |
-20°C | |
Liquid |
Western Blot | |
0.82 mg/mL | |
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35889, P55196, Q9QZQ1 | |
Afdn | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |