Learn More
AG 18, Tocris Bioscience™
EGFR/PDGFR-kinase inhibitor
Marke:  Tocris (Bio-Techne) 049350
Weitere Details : Netto-Gewicht : 0.01000kg
Menge. 50mg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417733
  • 525.22 € / 50 Milligramm
Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Inhibitor of epidermal growth factor receptor (EGFR) and platelet-derived growth factor receptor (PDGFR) kinase (IC 50 values are 35 and 25 μM respectively). Inhibits EGF-stimulated cell proliferation. Also acts as a mitochondrial uncoupler that alters phosphorylation-dependent cell signaling.
Spezifikation
Chemische Identifikatoren
118409-57-7
186.17
tyrphostin 23, tyrphostin a23, 3,4-dihydroxybenzylidene malononitrile, tyrphostin ag18, tyrphostin rg50810, unii-rv0gcd31oj, 2-3,4-dihydroxyphenyl methylidene propanedinitrile, 2-3,4-dihydroxybenzylidene malononitrile, alpha-cyano-3,4-dihydroxy cinnamonitrile, propanedinitrile,2-3,4-dihydroxyphenyl methylene
2-[(3,4-dihydroxyphenyl)methylidene]propanedinitrile
C10H6N2O2
VTJXFTPMFYAJJU-UHFFFAOYSA-N
2052
C1=CC(=C(C=C1C=C(C#N)C#N)O)O
Spezifikation
AG 18
118409-57-7
tyrphostin 23, tyrphostin a23, 3,4-dihydroxybenzylidene malononitrile, tyrphostin ag18, tyrphostin rg50810, unii-rv0gcd31oj, 2-3,4-dihydroxyphenyl methylidene propanedinitrile, 2-3,4-dihydroxybenzylidene malononitrile, alpha-cyano-3,4-dihydroxy cinnamonitrile, propanedinitrile,2-3,4-dihydroxyphenyl methylene
C1=CC(=C(C=C1C=C(C#N)C#N)O)O
186.17
186.17
50mg
C10H6N2O2
VTJXFTPMFYAJJU-UHFFFAOYSA-N
2-[(3,4-dihydroxyphenyl)methylidene]propanedinitrile
2052
>98%
Videos