missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Aggrecan G1-IGD-G2 Domains Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
N.V.
Spezifikation
Antigen | Aggrecan |
---|---|
Klon | 179509 |
Konzentration | LYOPH |
Verdünnung | Western Blot 1 μg/mL, Immunohistochemistry 5-25 μg/mL, Immunoprecipitation 25 μg/mL |
Anwendungen | Western Blot, Immunohistochemistry, Immunoprecipitation |
Beschreibung
Aggrecan Monoclonal specifically detects Aggrecan in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Spezifikation
Aggrecan | |
LYOPH | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
NP_037359 | |
176 | |
Mouse myeloma cell line NS0-derived recombinant human Aggrecan Val20-Gly675 Accession # NP_037359 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Aggrecan G1-IGD-G2 Domains in Western blots and direct ELISAs. |
179509 | |
Western Blot 1 μg/mL, Immunohistochemistry 5-25 μg/mL, Immunoprecipitation 25 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACAN, AGC1, AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16, MSK16AGCAN, SEDK | |
ACAN | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only