missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AgRP/ART Antibody, R&D Systems™
Mouse Monoclonal Antibody
N.V.
Spezifikation
Antigen | AgRP/ART |
---|---|
Klon | 82815 |
Verdünnung | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Anwendungen | ELISA |
Klassifikation | Monoclonal |
Beschreibung
AgRP/ART Monoclonal specifically detects AgRP/ART in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Spezifikation
AgRP/ART | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
agouti related protein homolog (mouse), agouti-related protein, Agrt, AGRTMGC118963, ART, ARTagouti (mouse) related protein, ASIP2 | |
AGRP | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
82815 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
O00253 | |
181 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human AgRP Ala21-Thr132 Accession # O00253 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human AgRP in ELISAs and Western blots. In Western blots, this antibody shows 100% cross-reactivity with recombinant mouse AgRP. |
For Research Use Only