missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHCYL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10658-3-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
AHCYL1 belongs to the adenosylhomocysteinase family. The protein, an inositol 1, 4, 5-trisphosphate receptor-binding protein, specifically binds to and activates pancreas-type Na+/HCO3- cotransporter 1 (pNBC1). The regulation through AHCYL1 enables NBC1 variants to have different physiological roles.Spezifikation
AHCYL1 | |
Polyclonal | |
Unconjugated | |
AHCYL1 | |
AdoHcyase 2, AHCYL1, DCAL, IRBIT, PRO0233, XPVKONA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10768, 229709, 362013 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B5DFN2, O43865, Q80SW1 | |
Ahcyl1 | |
AHCYL1 Fusion Protein Ag1017 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |