missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15618-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes an enzyme which reversibly catalyzes the phosphorolysis of purine nucleosides. The enzyme is trimeric, containing three identical subunits. Mutations which result in nucleoside phosphorylase deficiency result in defective T-cell (cell-mediated) immunity but can also affect B-cell immunity and antibody responses. Neurologic disorders may also be apparent in patients with immune defects. A known polymorphism at aa position 51 that does not affect enzyme activity has been described. A pseudogene has been identified on chromosome 2.Spezifikation
AKAP1 | |
Polyclonal | |
Unconjugated | |
AKAP1 | |
AKAP, AKAP 149, AKAP1, AKAP121, AKAP149, AKAP84, D AKAP 1, D AKAP1, PRKA1, S AKAP84, SAKAP84 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8165 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q92667 | |
AKAP1 | |
AKAP1 Fusion Protein Ag8037 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |