missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlaRS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17394-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The human alanyl-tRNA synthetase (AARS) belongs to a family of tRNA synthases,of the class II enzymes.Class II tRNA synthases evolved early in evolution and are highly conserved.This is reflected by the fact that 498 of the 968-residue polypeptide human AARS shares 41% identity witht the E.coli protein.tRNA synthases are the enzymes that interpret the RNA code and attach specific aminoacids to the tRNAs that contain the cognate trinucleotide anticodons.They consist of a catalytic domain which interacts with the amino acid acceptor-T psi C helix of the tRNA, and a second domain which interacts with the rest of the tRNA structure.Spezifikation
AlaRS | |
Polyclonal | |
Unconjugated | |
AARS | |
AARS, Alanine tRNA ligase, alanyl tRNA synthetase, AlaRS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16, 234734, 292023 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P49588, P50475, Q8BGQ7 | |
Aars | |
AlaRS Fusion Protein Ag11294 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |