Learn More
MP Biomedicals™ Albumin, Bovine, For IVD, Low Ash and Metals
Prepared by heat shock fractionation
Marke: MP Biomedicals™ 0216006910
Weitere Details : CAS : 9048-46-8 Netto-Gewicht : 0.01000kg
Beschreibung
- For IVD applications
- Low visible absorbance
- Purity: >98%
- Ash: <2%
- Metals: <20ppm
For best cell culture performance, see alternate Mfr. No. 180577
Spezifikation
ELISA, Microbial Culture | |
Bovine Albumin Fraction V | |
10g | |
2° to 8°C |
66.338 | |
6.5 to 7.5 | |
Bovine | |
Powder |