missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Albumin, Human, Purified Antigen Lyohphilized, FITC, 25mg
Marke: MP Biomedicals™ 0855883
Weitere Details : Netto-Gewicht : 0.00010kg
Spezifikation
Immunoassay | |
25mg | |
2° to 8°C |
Human Albumin | |
Human | |
Lyophilized |