missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH5A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17319-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Aldh5A1 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldehyde dehydrogenase enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. Aldh5A1 is a mitochondrial NAD(+)-dependent succinic semialdehyde dehydrogenase. A deficiency of this enzyme, known as 4-hydroxybutyricaciduria, results in a disorder of the neurotransmitter 4-aminobutyric acid (GABA). Symptoms usually include static encephalopathy, associated with developmental delays, hypotonia, ataxia, speech defects, and seizures. At least two isoforms of Aldh5A1 are known to exist.Spezifikation
ALDH5A1 | |
Polyclonal | |
Unconjugated | |
ALDH5A1 | |
ALDH5A1, SSADH, SSDH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
214579, 291133, 7915 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P51649, P51650, Q8BWF0 | |
Aldh5a1 | |
ALDH5A1 Fusion Protein Ag11293 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |