Learn More
ALK [L1196M] Recombinant Human Protein, Invitrogen™
Human ALK L1196M, internal fragment, amino acids L1066-S1437 (as in NCBI/Protein entry NP_004295.2), L1196M mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.
Marke: Invitrogen PV6166
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Spezifikation
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
70.5kDa | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
aa 1066-1437 | |
Recombinant |
50mM HEPES pH 7.5, 100mM NaCl, 5mM DTT, 15mM reduced glutathione, 20% glycerol | |
10μg | |
ALK, CD246, NBLST3 | |
Human | |
GST Tag, His Tag |