missing translation for 'onlineSavingsMsg'
Learn More

Allylcyclopentan, 98 %, Thermo Scientific Chemicals

Catalog Number p-7024953
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3524-75-2
C8H14
110.2
MFCD00001390
NHIDGVQVYHCGEK-UHFFFAOYSA-N
allylcyclopentane, 3-cyclopentylpropene, cyclopentane, 2-propenyl, cyclopentane, allyl, 3-cyclopentyl-1-propene, allyl cyclopentane, 2-propenylcyclopentane, 1-propene, 3-cyclopentyl, 3-cyclopentylprop-1-ene
77059
Prop-2-enylcyclopentan
C=CCC1CCCC1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3524-75-2
C8H14
110.2
MFCD00001390
NHIDGVQVYHCGEK-UHFFFAOYSA-N
allylcyclopentane, 3-cyclopentylpropene, cyclopentane, 2-propenyl, cyclopentane, allyl, 3-cyclopentyl-1-propene, allyl cyclopentane, 2-propenylcyclopentane, 1-propene, 3-cyclopentyl, 3-cyclopentylprop-1-ene
77059
Prop-2-enylcyclopentan
C=CCC1CCCC1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Allylcyclopentane
Schmelzpunkt -111°C
Dichte 0.793
Siedepunkt 125°C to 127°C
Flammpunkt 12°C (53°F)
Brechungsindex 1.441
Menge 1 g
UN-Nummer UN3295
Beilstein 2036419
Formelmasse 110.2
Reinheit (%) 98%
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'