missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methyl-DL-phenylalanin, 98 %, Thermo Scientific™

Catalog Number p-7028040
Click to view available options
Menge:
0.25 g
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1132-26-9
C10H13NO2
179.22
MFCD00010512
IRZQDMYEJPNDEN-UHFFFAOYNA-N
alpha-methyl-dl-phenylalanine, 2-amino-2-methyl-3-phenylpropionic acid, alpha-methylphenylalanine, h-dl-me phe-oh, h-alpha-me-dl-phe-oh, +/--2-amino-2-methyl-3-phenylpropionic acid, phenylalanine, alpha-methyl, 2-amino-2-methyl-3-phenyl-propanoic acid, h-me-dl-phe-oh, 2-methylphenylalanine #
108055
2-amino-2-methyl-3-Phenylpropansäure
CC(C(N)C(O)=O)C1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1132-26-9
C10H13NO2
179.22
MFCD00010512
IRZQDMYEJPNDEN-UHFFFAOYNA-N
alpha-methyl-dl-phenylalanine, 2-amino-2-methyl-3-phenylpropionic acid, alpha-methylphenylalanine, h-dl-me phe-oh, h-alpha-me-dl-phe-oh, +/--2-amino-2-methyl-3-phenylpropionic acid, phenylalanine, alpha-methyl, 2-amino-2-methyl-3-phenyl-propanoic acid, h-me-dl-phe-oh, 2-methylphenylalanine #
108055
2-amino-2-methyl-3-Phenylpropansäure
CC(C(N)C(O)=O)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material alpha-Methyl-DL-phenylalanine
Schmelzpunkt 293°C to 294°C (decomposition)
Formelmasse 179.22
Reinheit (%) 98%
Menge 0.25 g
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'