missing translation for 'onlineSavingsMsg'
Learn More

Aluminiumkugel, 6.35 mm (0.25 Zoll) im Durchmesser x 6.35 mm (0.25 Zoll) Länge, Puratronic™, 99.999 % (Metallbasis), Thermo Scientific Chemicals

Catalog Number p-7051957
Click to view available options
Menge:
10 g
50 g
250 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
Aluminium
[Al]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
Aluminium
[Al]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Aluminum slug
Namenshinweis 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
Hinweise zum Prozentgehalt (Metallbasis)
Menge 10 g
Reinheit (%) 99.999%
Güte Puratronic™
Geruch Odorless
Gewicht ≈0.54g/slug
Form Slug

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'