Learn More
Invitrogen™ Ambion™ MAXIscript™ T7 Transcription Kit
Spezifikation
Spezifikation
| Inhalt und Lagerung | T7 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, and pTRI-b-actin are all stored at -20°C. Nuclease-free Water may be stored at any temperature. |
| Zur Verwendung mit (Anwendung) | DNA and RNA Purification and Analysis, Gene Expression Analysis and Genotyping, in vitro Transcription, Northern Blotting, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis and Blotting, Southern Blotting |
| Enthält | T7 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, pTRI-b-actin, and Nuclease-free Water |
| Anzahl Reaktionen | 30 |
| Produkttyp | Kit with manual |
| Menge | 30 Reaktionen |