missing translation for 'onlineSavingsMsg'
Learn More

Ambrisentan, Thermo Scientific Chemicals

Catalog Number p-7214478
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
177036-94-1
C22H22N2O4
378.43
OUJTZYPIHDYQMC-UHFFFAOYNA-N
2-[(4,6-dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropanoic acid
COC(C(OC1=NC(C)=CC(C)=N1)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
177036-94-1
C22H22N2O4
378.43
OUJTZYPIHDYQMC-UHFFFAOYNA-N
2-[(4,6-dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropanoic acid
COC(C(OC1=NC(C)=CC(C)=N1)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ambrisentan
Schmelzpunkt >150.0°C
Menge 250 mg
Löslichkeitsinformationen Solubility aq. soln.: insoluble.
Formelmasse 378.42

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'