missing translation for 'onlineSavingsMsg'
Learn More

Amidosulfonsäure, ACS, 99.3-100.3 % (Assay-Trockenbasis), Thermo Scientific Chemicals

Catalog Number p-7050007
Click to view available options
Menge:
100 g
500 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5329-14-6
H3NO3S
97.088
MFCD00011603
IIACRCGMVDHOTQ-UHFFFAOYSA-N
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
5987
CHEBI:9330
Sulfamidsäure
NS(=O)(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5329-14-6
H3NO3S
97.088
MFCD00011603
IIACRCGMVDHOTQ-UHFFFAOYSA-N
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
5987
CHEBI:9330
Sulfamidsäure
NS(=O)(=O)O

Amidosulfonic acid is widely utilized as a green catalyst for the preparation of amide from ketoxime. It finds an application as a titrant in the determination of the burette injection volume and chemical calibration factor. It is an efficient catalyst in the synthesis of polyhydroquinoline derivatives by Hantzsch condensation reaction, deazaoxaflavin, fatty acid methyl and ethyl esters and in the isolation of 5-hydroxymethylfurfural (HMF) from bamboo fiber. It is involved in the determination of silicates in water samples. It acts as a neutralizing agent during the determination of paracetamol by Glynn and Kendal colorimetric method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Amidosulfonic acid
Schmelzpunkt ∼205°C (decomposition)
Dichte 2.12
Prozentgehaltsbereich 99.3 to 100.3% (Assay dried basis)
Hinweise zum Prozentgehalt (Assay-Trockenbasis)
Menge 100 g
Physikalische Form Kristallin
UN-Nummer UN2967
Merck Index 148921
Löslichkeitsinformationen Slightly soluble in water,dimethyl fluoride and methanol. Insoluble in hydrocarbons.
Formelmasse 97.09
Reinheit (%) 99.3 to 100.3%
Güte ACS
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'