missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bismuth citrate, Bi 48-52%, water ca 2%

Product Code. 11468846
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
Product Code. Quantity unitSize
11468846 1000 g 1000g
11478846 50 g 50g
11488846 250 g 250g
3 options
This item is not returnable. View return policy

Product Code. 11468846

Brand: Thermo Scientific Alfa Aesar A17003.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25530-63-6 | C6H11BiNO7 | 418.134 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25530-63-6
Molecular Formula C6H11BiNO7
Molecular Weight (g/mol) 418.134
MDL Number MFCD00036420
InChI Key JDINTLWFPXWXCT-UHFFFAOYSA-N
Synonym ammonium bismuth citrate
PubChem CID 87101692
IUPAC Name azane;bismuth;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Bi]

Specifications

Quantity 1000 g
Formula Weight 433.12
Assay Bi 48 to 52%, water ≈2%
Chemical Name or Material Ammonium bismuth citrate
compliance-icons
Product Identifier
  • Ammonium bismuth citrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P363-Wash contaminated clothing before reuse.
  • P321-Specific treatment (see on this label).
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.