Learn More
Ammonium Hexafluorophosphate, TRC
Product Code. 30340795
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340795 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30340795 Supplier Toronto Research Chemicals Supplier No. TRCA6340251G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ammonium Hexafluorophosphate
CAS 16941-11-0
Quantity 1 g
Molecular Formula F6H4NP
Synonym Hexafluoro-phosphate(1-) Ammonium, Ammonium Hexafluorophosphate, Ammonium Hexafluorophosphate(1-), NSC 403554, NSC 407930
InChI Formula InChI=1S/F6P.H3N/c1-7(2,3,4,5)6;/h;1H3/q-1;/p+1
SMILES [NH4+].F[P-](F)(F)(F)(F)F
IUPAC Name azanium;hexafluorophosphate
Molecular Weight (g/mol) 163
Formula Weight 162.9986