Learn More
Ammonium L-(+)-tartrat, 98 %, Thermo Scientific Chemicals
Beschreibung
Ammonium L-(+)-tartrate finds application in the textile and pharmaceutical industries. It is also used to study protein structural analysis, biochemicals and reagents, biological buffers, chelators and proteomics. It acts as a nitrogen source in biological applications and cell culture media. Further, it is used in the study of crystal optimization research and preliminary diffraction data analysis of the Smad1 MH1 domain bonded to a palindromic SBE DNA element.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Chemischer Name oder Material | Ammonium L-(+)-tartrate |
Farbe | Weiß |
Physikalische Form | Kristallin |
Menge | 50 g |
Beilstein | 6120352 |
Löslichkeitsinformationen | Soluble in water and alcohol. |
Formelmasse | 184.15 |
Reinheit (%) | 98% |
Geruch | Odorless |
Empfindlichkeit | Hygroscopic |
Mehr anzeigen |
RUO – Research Use Only