missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammoniumperchlorat-Lösung, Honeywell
Click to view available options
Menge:
1 l
2.5 l
Spezifikation
Chemischer Name oder Material | Ammoniumperchloratlösung |
Menge | 1 l |
Strukturformel | NH4ClO4 |
UN-Nummer | 3211 |
Formelmasse | 117.49 mol |
Güte | Reagenz |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'